Dowex® 1X8 (Cl) 200-400 mesh ion-exchange resin

AC20301-5000 AC20301-1000
200018-982EA 151.15 USD
200018-982 200006-404
Dowex® 1X8 (Cl) 200-400 mesh ion-exchange resin
Dowex® 1X8 (Cl)
Storage Temperature: Ambient
MDL Number: MFCD00132718
CAS Number: 12627-85-9

Order Now

Specification Test Results

Appearance White to yellow Moist beads
Particle size ≤15 % (on 100 mesh, cumulative)
Particle size ≤15 % (through 325 mesh)
Wet volume capacity ≥1.2 meq/ml (Cl form)
Water retention capacity 39 to 45 % (Cl form)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR