Amberlite® XAD-7 HP

AC20224-5000 AC20224-1000
200018-138EA 47.08 USD
200018-138 200011-170
Amberlite® XAD-7 HP
Amberlite® XAD-7 HP
Storage Temperature: Ambient
MDL Number: MFCD00132705
CAS Number: 37380-43-1

Order Now

Specification Test Results

Appearance White to pink-beige Beads
Particle size ≤7.0 % (< 0.300 mm)
Particle size ≤8.0 % (> 1.180 mm)
Harmonic mean size 0.430 to 0.690 mm
Uniformity coefficient ≤2.0
TOC-AFNOR test ≤1 ppm (at 5V/V)
TOC-AFNOR test ≤10 ppm (total)
Moisture holding capacity 61.0 to 69.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR