Amberlite® CG-50 ion-exchange resin Type 1

AC20211-1000 AC20211-5000 AC20211-0250
200011-158EA 81.82 USD
200011-158 103467-280 103467-248
Amberlite® CG-50 ion-exchange resin Type 1
Amberlite® CG-50
Storage Temperature: Ambient
MDL Number: MFCD00132693
CAS Number: 9042-11-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Particle size ≤10 % (< 75 µm)
Particle size ≤10 % (> 150 µm)
Moisture ≤10 %
Total weight capacity ≥10 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR