D-(-)-α-(4-Hydroxyphenyl)glycine 98+%

Warning

Synonyms: H-D-Phg(4-OH)-OH(-)-4-Hydroxy-D-phenylglycine(R)-(-)-2-(4-Hydroxyphenyl)glycineD-(-)-p-HydroxyphenylglycineD(-)-4-Hydroxyphenylglycine

AC20132-1000 AC20132-0250
200024-900EA 99.6 USD
200024-900 200030-848
D-(-)-α-(4-Hydroxyphenyl)glycine 98+%
D(-)-α-(4-Hydroxyphenyl)glycine
Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: 240 °C (dec.)
Storage Temperature: Ambient
EINECS: 245-247-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Specific optical rotation -155° to -161° (20 °C, 589 nm) (c=1, 1N HCl)
Titration with HClO4 ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR