Benzylammonium chloride 99%

Warning

Synonyms: Benzylamine hydrochloride

AC20054-0050 AC20054-1000
200018-270EA 103 USD
200018-270 200018-272
Benzylammonium chloride 99%
Benzylammonium chloride
Formula: C₇H₁₀ClN
MW: 143.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012852
CAS Number: 3287-99-8
EINECS: 221-943-6

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 259 to 264 °C
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR