4-Iodo-1H-pyrazole 98%

Warning

Synonyms: 4-Iodopyrazole

AC20037-0050 AC20037-0250
102775-522EA 95.5 USD
102775-522 102777-140
4-Iodo-1H-pyrazole 98%
4-Iodo-1H-pyrazole
Formula: C₃H₃IN₂
MW: 193.97 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005244
CAS Number: 3469-69-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 107 to 112 °C
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR