Sodium triethylborohydride 1 M in THF, AcroSeal™

Danger

Synonyms: Borate(1-), triethylhydro-, sodium, (T-4)-sodium triethylhydroborateSodium triethylborohydride

AC20003-8000 AC20003-1000
102777-522EA 745.5 USD
102777-522 103526-544
Sodium triethylborohydride 1 M in THF, AcroSeal™
Sodium triethylborohydride
Formula: C₆H₁₆BNa
MW: 121.99 g/mol
Density: 0.89 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011704
CAS Number: 17979-81-6
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Colorless Solution
Titration 0.95 to 1.10 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR