Benzeneseleninic anhydride 98%

Danger

Synonyms: Benzeneseleninic acid anhydride

AC19971-0250
102777-338EA 388 USD
102777-338
Benzeneseleninic anhydride 98%
Benzeneseleninic anhydride
Formula: C₁₂H₁₀O₃Se₂
MW: 360.13 g/mol
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00001991
CAS Number: 17697-12-0
EINECS: 241-701-3
UN: 3283
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to yellow to light beige Crystalline powder or needles
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR