Sulfamide 98+%

Warning

Synonyms: Sulphuric diamideSulfuric diamide

AC19902-0250 AC19902-0050
200020-768EA 53.9 USD
200020-768 200020-766
Sulfamide 98+%
Sulfamide
Formula: H₄N₂O₂S
MW: 96.11 g/mol
Melting Pt: 87…91 °C
Density: 1.61 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011606
CAS Number: 7803-58-9
EINECS: 232-262-9

Order Now

Specification Test Results

Appearance White to practically white Crystalline powder (Free from visible impurities)
Infrared spectrum Conforms
Melting point 89.0 to 93.0 °C
Loss on drying ≤0.5 % (50 °C, 2 hrs) (vacuum)
Elemental Analysis ≥98.0 %
Related substances (TLC) Passes test
pH 5.0 to 7.0 (10 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR