5-Butylpicolinic acid 99%

Warning

Synonyms: Fusaric acid

AC19896-2500 AC19896-0010
200012-550EA 223 USD
200012-550 102774-978
5-Butylpicolinic acid 99%
5-Butylpicolinic acid
Formula: C₁₀H₁₃NO₂
Melting Pt: 97…101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006298
CAS Number: 536-69-6
EINECS: 208-643-0
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 97 to 101 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR