L-allo-Threonine 99%

Synonyms: H-allo-Thr-OHL(+)-allo-ThreonineAllo-Thr-OH

AC19854-1000 AC19854-2500
103509-510EA 94.2 USD
103509-510 200019-450
L-allo-Threonine 99%
L-allo-Threonine
Formula: C₄H₉NO₃
MW: 119.12 g/mol
Melting Pt: 256 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064268
CAS Number: 28954-12-3
EINECS: 249-327-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Specific optical rotation +8° to +10° (20 °C, 589 nm) (c=2, water)
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR