Ethyl-N-hydroxyacetimidate 97%

Warning

Synonyms: Ethyl acetohydroximate

AC19750-0500 AC19750-0100
200019-146EA 46.8 USD
200019-146 200037-404
Ethyl-N-hydroxyacetimidate 97%
Ethyl N-hydroxyacetimidate
Formula: C₄H₉NO₂
MW: 103.12
Boiling Pt: 55…58 °C (6 mmHg)
Melting Pt: 23…25 °C
Flash Pt: 77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002114
EINECS: 234-165-7

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4320 to 1.4350 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR