Osmic acid 2.5% (w/w) in tert-butanol stabilized

Danger

Synonyms: Osmium TetroxideOsmium tetraoxideOsmic Acid AnhydrideOsmium (VIII) oxide

AC19745-0010 AC19745-0050 AC19745-0250
102774-260EA 57.82 USD
102774-260 200002-176 102777-142
Osmic acid 2.5% (w/w) in tert-butanol stabilized
Osmic acid
Formula: O₄Os
MW: 254.23 g/mol
Flash Pt: 4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011150
CAS Number: 20816-12-0
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Appearance White to yellow Fused mass or solution
Stabilizer tert-Butyl hydroperoxide
Strength ≥2.5 wt%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR