Thallium(I) sulfate 99.9+% (trace metals basis)

Danger

Synonyms: Thallic sulfateThallic sulphateDithallium sulfateDithallium sulphate

AC19694-2500 AC19694-0500
103526-498EA 452 USD
103526-498 102775-896
Thallium(I) sulfate 99.9+% (trace metals basis)
Dithallium sulphate
Formula: O₄STl₂
MW: 504.83 g/mol
Melting Pt: 632 °C
Density: 6.77 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011278
CAS Number: 7446-18-6
EINECS: 231-201-3
UN: 1707
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Assay 80.0 to 84.0 % (Tl)
Total trace metal impurities ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR