6-Methoxy-8-nitroquinoline 99%

AC19560-0010
200010-816EA 53.5 USD
200010-816
6-Methoxy-8-nitroquinoline 99%
6-Methoxy-8-nitroquinoline
Formula: C₁₀H₈N₂O₃
MW: 204.1800 g/mol
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00006802
CAS Number: 85-81-4
EINECS: 201-633-7

Order Now

Specification Test Results

Appearance Yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 158 to 162 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR