1-Methylcyclopropanecarboxylic acid 98%

Danger

Synonyms: 1-methylcyclopropane-1-carboxylic acid

AC19496-0050 AC19496-0010
102776-178EA 190 USD
102776-178 102774-520
1-Methylcyclopropanecarboxylic acid 98%
1-Methylcyclopropanecarboxylic acid
Formula: C₅H₈O₂
MW: 100.12 g/mol
Melting Pt: 34…36 °C
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00001290
CAS Number: 6914-76-7
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light yellow Low melting solid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR