Sodium borodeuteride (98% D) for NMR spectroscopy

Danger

AC19495-0050
102776-748EA 270 USD
102776-748
Sodium borodeuteride (98% D) for NMR spectroscopy
Sodium borodeuteride
Formula: D₄BNa
MW: 41.857 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
EINECS: 239-764-7
UN: 1426
ADR: 4.3,I

Order Now

Specification Test Results

Appearance White to off-white to light grey Powder and/or chunks
D-Enrichment ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR