1-Acetylpiperidine-4-carboxylic acid 99%

Warning

AC19463-0050
200063-470EA 38 USD
200063-470
1-Acetylpiperidine-4-carboxylic acid 99%
1-Acetylpiperidine-4-carboxylic acid
Formula: C₈H₁₃NO₃
MW: 171.19 g/mol
Melting Pt: 180…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00023706
CAS Number: 25503-90-6
EINECS: 247-049-6

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 182 to 185 °C
Titration with TBAH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR