Janus green B, pure, high purity biological stain

AC19168-0250 AC19168-0050
103511-166EA 183 USD
103511-166 200012-904
Janus green B, pure, high purity biological stain
Janus green B
Formula: C₃₀H₃₁ClN₆
Storage Temperature: Ambient
MDL Number: MFCD00011758
CAS Number: 2869-83-2

Order Now

Specification Test Results

Appearance Dark green to dark brown to black Powder
Infrared spectrum Conforms
Dye content ≥50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR