Lithium hydride 97%, powder (<100 µm), pure

Danger

AC19119-0100 AC19119-1000
102774-692EA 23.1 USD
102774-692 103531-730
Lithium hydride 97%, powder (<100 µm), pure
Lithium hydride
Formula: HLi
Melting Pt: 680 °C
Density: 0.82 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011074
CAS Number: 7580-67-8
EINECS: 231-484-3
UN: 1414
ADR: 4.3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR