Fluorescein diacetate 97%, pure

AC19166-0050 AC19166-0250
103511-160EA 212 USD
103511-160 200019-202
Fluorescein diacetate 97%, pure
Fluorescein diacetate
Formula: C₂₄H₁₆O₇
Melting Pt: 200…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00005062
CAS Number: 596-09-8
EINECS: 209-877-6

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 200 to 205 °C
Sulfated ash ≤0.1 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR