Hexafluorophosphoric acid 60% (w/w) in water

Danger

AC19111-0250 AC19111-5000
200057-736EA 21.35 USD
200057-736 200057-738
Hexafluorophosphoric acid 60% (w/w) in water
Hexafluorophosphoric acid
Formula: HF₆P
Density: 1.75 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011350
CAS Number: 16940-81-1
EINECS: 241-006-5
UN: 2922
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR