N-Benzylbenzamide 99%

Warning

AC19117-0250 AC19117-0050
200013-576EA 87.8 USD
200013-576 200032-144
N-Benzylbenzamide 99%
N-Benzylbenzamide
Formula: C₁₄H₁₃NO
MW: 211.2591 g/mol
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00003070
CAS Number: 1485-70-7
EINECS: 216-063-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR