2,3-Dihydrofuran 98+%

Danger

AC19044-1000 AC19044-5000
200016-038EA 214 USD
200016-038 200008-918
2,3-Dihydrofuran 98+%
2,3-Dihydrofuran
Formula: C₄H₆O
Density: 0.927 g/cm³
Flash Pt: –24 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003205
CAS Number: 1191-99-7
EINECS: 214-747-7
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR