Ethyl-5-methyl-1H-imidazole-4-carboxylate 98%

Synonyms: Ethyl 5-methyl-1H-imidazole-4-carboxylate4-Methyl-1H-imidazole-5-carboxylic acid ethyl esterEthyl 4-methyl-5-imidazolecarboxylateEthyl 4-Methyl-1H-imidazole-5-carboxylate

AC19034-0500 AC19034-0100
200049-360EA 484 USD
200049-360 200025-312
Ethyl-5-methyl-1H-imidazole-4-carboxylate 98%
Ethyl-5-methyl-1H-imidazole-4-carboxylate

Appearance: Off-white to tan Crystalline powder or crystals

Formula: C₇H₁₀N₂O₂
MW: 154.1665 g/mol
Melting Pt: 204…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00005199
CAS Number: 51605-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR