Acid Red 18 (Food Red No. 102), pure

Synonyms: Sodium 7-hydroxy-8-((4-sulfonatonaphthalen-1-yl)diazenyl)naphthalene-1,3-disulfonateCochineal Red APonceau 4RCochenille redScarlet 3RSodium 7-hydroxy-8-((4-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-1,3-disulphonateNew Coccine

AC19023-1000 AC19023-0250
200026-134EA 27 USD
200026-134 103511-150
Acid Red 18 (Food Red No. 102), pure
Acid Red 18 (Food Red No. 102)
Formula: C₂₀H₁₄N₂O₁₀S₃
MW: 604.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004084
CAS Number: 2611-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR