Acid green 50, pure

Warning

Synonyms: Green SLissamine Green B

AC19010-0250
200019-506EA 46 USD
200019-506
Acid green 50, pure
Acid green 50

Appearance: Dark brown to black Powder

Formula: C₂₇H₂₆N₂O₇S₂
MW: 576.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012117
CAS Number: 3087-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR