Sodium borohydride 98+%, powder
|
Danger
|
Synonyms:
Sodium tetrahydridoborate
, Sodium tetrahydroborate
AC18930-0025
103526-402EA
777
USD
103526-402
Sodium borohydride 98+%, powder
Sodium borohydride
Formula:
NaBH₄ MW: 37.83 g/mol Melting Pt: 360 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003518 CAS Number: 16940-66-2 EINECS: 241-004-4 UN: 1426 ADR: 4.3,I Merck Index: 12,08735 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...