Sodium bis(2-methoxyethoxy)aluminium hydride ∼3.5 M in toluene 70% (w/w)

Danger

Synonyms: SBAHSodium dihydrobis(2-methoxyethoxy)aluminateSodium bis(2-methoxyethoxy)aluminum dihydride

AC18790-0500
102774-086EA 40.3 USD
102774-086
Sodium bis(2-methoxyethoxy)aluminium hydride ∼3.5 M in toluene 70% (w/w)
Sodium bis(2-methoxyethoxy)aluminium hydride
Formula: C₆H₁₆AlNaO₄
MW: 202.16 g/mol
Density: 1.036 g/cm³
Flash Pt: 4 °C
Storage Temperature: Ambient
EINECS: 245-178-2
UN: 3399
ADR: 4.3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR