Thiophene-2-carbonyl chloride 98%

Danger

Synonyms: Thiophene-2-carboxylic acid chloride2-Thiophenecarbonyl chloride

AC18777-0250 AC18777-1000
200016-766EA 33.9 USD
200016-766 200028-960
Thiophene-2-carbonyl chloride 98%
Thiophene-2-carbonyl chloride
Formula: C₅H₃ClOS
MW: 146.6 g/mol
Density: 1.37 g/cm³
Flash Pt: 90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005428
CAS Number: 5271-67-0
EINECS: 226-092-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR