Ethylenediammonium dichloride 98+%

Warning

Synonyms: Ethylenediamine dihydrochloride

AC18765-1000 AC18765-5000 AC18765-0025
103511-118EA 429 USD
103511-118 200006-522 200006-520
Ethylenediammonium dichloride 98+%
Ethylenediammonium dichloride
Formula: C₂H₁₀Cl₂N₂
MW: 133.02 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00012524
CAS Number: 333-18-6
EINECS: 206-369-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR