Ethyl isothiocyanatoformate 95%

Danger

Synonyms: Ethoxycarbonyl isothiocyanate

AC18785-0250 AC18785-0050
102774-914EA 77 USD
102774-914 102776-570
Ethyl isothiocyanatoformate 95%
Ethyl isothiocyanatoformate
Formula: C₄H₅NO₂S
MW: 131.16 g/mol
Boiling Pt: 56 °C (18 mmHg)
Density: 1.16 g/cm³
Flash Pt: 50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004814
CAS Number: 16182-04-0
UN: 2929
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR