2,5-Dihydroxy-p-benzoquinone 98%

Warning

Synonyms: 2,5-Dihydroxy-1,4-benzoquinone

AC18746-1000 AC18746-0250
200022-240EA 188 USD
200022-240 200022-242
2,5-Dihydroxy-p-benzoquinone 98%
2,5-Dihydroxy-p-benzoquinone
Formula: C₆H₄O₄
MW: 140.1 g/mol
Melting Pt: 220 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00001598
CAS Number: 615-94-1
EINECS: 210-454-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR