6-Methoxytryptamine 99%

Danger

AC18745-0010 AC18745-2500 AC18745-0050 AC18745-0500
102774-272EA 33.7 USD
102774-272 102777-314 103531-716 200005-584
6-Methoxytryptamine 99%
6-Methoxytryptamine
Formula: C₁₁H₁₄N₂O
Melting Pt: 143…146 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005663
EINECS: 222-778-2
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR