tert-Butylhydrazine hydrochloride 98%

Warning

AC18708-5000 AC18708-1000
200050-906EA 501.4 USD
200050-906 200020-812
tert-Butylhydrazine hydrochloride 98%
tert-Butylhydrazine hydrochloride
Formula: C₄H₁₃ClN₂
MW: 124.61 g/mol
Melting Pt: 190…194 °C
Storage Temperature: Ambient
EINECS: 231-001-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR