Ethyl-3-piperidinecarboxylate 97%

Danger

Synonyms: Ethyl piperidine-3-carboxylateEthyl nipecotate

AC18662-0250 AC18662-1000 AC18662-5000
200019-148EA 42.09 USD
200019-148 200012-424 102777-240
Ethyl-3-piperidinecarboxylate 97%
Ethyl 3-piperidinecarboxylate
Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Boiling Pt: 102…104 °C (9hPa)
Density: 1.012 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00005991
CAS Number: 5006-62-2
EINECS: 225-681-3
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR