3-(Trifluoromethyl)phenylacetone 97%

Warning

Synonyms: 1-[3-(Trifluoromethyl)phenyl]-2-propanone

AC18616-0100
200049-810EA 110.9 USD
200049-810
3-(Trifluoromethyl)phenylacetone 97%
3-(Trifluoromethyl)phenylacetone
Formula: C₁₀H₉F₃O
MW: 202.18 g/mol
Boiling Pt: 89…90 °C (.5 mmHg)
Density: 1.204 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
EINECS: 244-652-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR