Tryptamine 98%

Danger

Synonyms: 2-(1H-indol-3-yl)ethan-1-amine

AC18612-0100 AC18612-0500
200008-030EA 45.5 USD
200008-030 200008-032
Tryptamine 98%
Tryptamine
Formula: C₁₀H₁₂N₂
Boiling Pt: 137 °C (.1 mmHg)
Melting Pt: 113…117 °C
Flash Pt: 185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005661
CAS Number: 61-54-1
EINECS: 200-510-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR