tert-Butylchlorodiphenylsilane 98%

Danger

Synonyms: tert-Butyldiphenylchlorosilanetert-Butyl-diphenylchlorosilane

AC18750-2500
102777-492EA 450 USD
102777-492
tert-Butylchlorodiphenylsilane 98%
tert-Butylchlorodiphenylsilane
Formula: C₁₆H₁₉ClSi
MW: 274.87 g/mol
Boiling Pt: 135 °C (.7 mmHg)
Density: 1.06 g/cm³
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00000497
CAS Number: 58479-61-1
EINECS: 261-282-0
UN: 2987
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR