6-Amino-1,4-benzodioxane 99%

Warning

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxin-6-amine1,4-Benzodioxan-6-amine

AC18578-0500 AC18578-0050 AC18578-2500
200027-682EA 35.4 USD
200027-682 200027-680 102777-614
6-Amino-1,4-benzodioxane 99%
6-Amino-1,4-benzodioxane
Formula: C₈H₉NO₂
MW: 151.17 g/mol
Density: 1.23 g/cm³
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00006824
CAS Number: 22013-33-8
EINECS: 244-718-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR