2-Mesitylenesulfonyl hydrazide 97%

Warning

Synonyms: 2,4,6-Trimethylbenzenesulfonyl Hydrazide2,4,6-Trimethylbenzenesulphonyl Hydrazide

AC18509-0250
200034-312EA 180.4 USD
200034-312
2-Mesitylenesulfonyl hydrazide 97%
2-Mesitylenesulfonyl hydrazide

Appearance: White to off-white Crystalline powder

Formula: C₉H₁₄N₂O₂S
MW: 214.28 g/mol
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00007585
CAS Number: 16182-15-3
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR