7-Aminocephalosporanic acid 95-102%

Danger

Synonyms: 7-ACA(6R,7R)-3-(Acetoxymethyl)-7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

AC18441-0050 AC18441-0010
200030-200EA 42.91 USD
200030-200 200017-916
7-Aminocephalosporanic acid 95-102%
7-Aminocephalosporanic acid

Appearance: Off-white to beige Crystalline powder

Formula: C₁₀H₁₂N₂O₅S
MW: 272.28 g/mol
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005177
CAS Number: 957-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR