Ethyl succinyl chloride 95%

Danger

Synonyms: Ethyl 4-chloro-4-oxobutanoate

AC18409-0250 AC18409-1000
200019-172EA 63.4 USD
200019-172 200031-348
Ethyl succinyl chloride 95%
Ethyl succinyl chloride
Formula: C₆H₉ClO₃
MW: 164.59 g/mol
Boiling Pt: 88…90 °C (11 mmHg)
Density: 1.1575 g/cm³
Flash Pt: 84 °C
Storage Temperature: Ambient
EINECS: 238-855-9
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR