4-Bromophenethylamine 98%

Danger

Synonyms: 2-(4-Bromophenyl)ethylamine

AC18309-0050 AC18309-0010 AC18309-0250
200050-384EA 53 USD
200050-384 200013-906 102777-732
4-Bromophenethylamine 98%
4-Bromophenethylamine
Formula: C₈H₁₀BrN
Boiling Pt: 63…72 °C (.2 mmHg)
Density: 1.29 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008189
EINECS: 277-636-2
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR