o-Cresolphthalein complexone, pure

Synonyms: Cresol phthalexonePhthalein complexoneo-CresolphthalexonXylenolphthalein-bis(iminodiacetic acid)Phthalein purpleMetal phthaleine

AC18244-0250 AC18244-0050
200026-276EA 165 USD
200026-276 200020-210
o-Cresolphthalein complexone, pure
o-Cresolphthalein complexone
Formula: C₃₂H₃₂N₂O₁₂
MW: 636.62 g/mol
Melting Pt: 181 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005911
CAS Number: 2411-89-4
EINECS: 219-318-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR