Bis(trimethylsilyl)acetylene 99%

Danger

AC18201-2500
103526-386EA 654.2 USD
103526-386
Bis(trimethylsilyl)acetylene 99%
Bis(trimethylsilyl)acetylene
Formula: C₈H₁₈Si₂
Boiling Pt: 136…137 °C (760 mmHg)
Melting Pt: 21…24 °C
Density: 0.75 g/cm³
Flash Pt: 2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008276
CAS Number: 14630-40-1
EINECS: 238-671-9
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR