Cyanamide 95% stabilized

Danger

AC18195-0250 AC18195-1000 AC18195-5000
102777-034EA 311 USD
102777-034 200011-792 200011-794
Cyanamide 95% stabilized
Cyanamide
Formula: CH₂N₂
MW: 42.04
Melting Pt: 40…47 °C
Flash Pt: 141 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007572
CAS Number: 420-04-2
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR