Pyridine hydrofluoride 65 - 70% approx. 30% Pyridine
|
Danger
|
Synonyms:
Olah's reagent
AC18007-0500
AC18007-0100
AC18007-1000
200057-846EA
109.85
USD
200057-846
200057-848
200057-850
Pyridine hydrofluoride 65 - 70% approx. 30% Pyridine
Pyridine hydrofluoride
Appearance: Clear light yellow to light brown Liquid
Formula:
C₅H₆FN MW: 99.108 g/mol Density: 1.1 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00012436 CAS Number: 32001-55-1 UN: 1790 ADR: 8,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...