tert-Butyl hydroperoxide 70% in water

Danger

Synonyms: TBHP

AC18034-0100 AC18034-0050 AC18034-5000
103524-134EA 352 USD
103524-134 200058-602 200058-606
tert-Butyl hydroperoxide 70% in water
tert-Butyl hydroperoxide
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 96 °C (dec.)
Melting Pt: –3 °C
Density: 0.937
Flash Pt: 43 °C(109 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002130
CAS Number: 75-91-2
EINECS: 200-915-7
UN: 3109
ADR: 5.2,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR