1,4-Dithiane-2,5-diol 96%

Danger

Synonyms: 2,5-Dihydroxy-1,4-dithianep-Dithiane-2,5-diol

AC17993-2500
200032-454EA 145 USD
200032-454
1,4-Dithiane-2,5-diol 96%
1,4-Dithiane-2,5-diol
Formula: C₄H₈O₂S₂
MW: 152.24 g/mol
Melting Pt: 130 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00006659
CAS Number: 40018-26-6
EINECS: 254-751-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR